Product Details

SNP ID
rs4633392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123011519 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTTCCAAAGCATTTTAATGTAG[C/G]TTGATATCTTTTTTTTTTTTTTTTT
Phenotype
MIM: 600301 MIM: 606238
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACADSB PubMed Links

Gene Details

Gene
ACADSB
Gene Name
acyl-CoA dehydrogenase, short/branched chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001609.3 Intron NP_001600.1
XM_017016194.1 Intron XP_016871683.1
Gene
IKZF5
Gene Name
IKAROS family zinc finger 5
There are no transcripts associated with this gene.

View Full Product Details