Product Details

SNP ID
rs4651137
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:183025165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTTCCAGAGAGAACAGACTGCC[C/T]CAAGCCATTCTTCTGTCAAGGGAGG
Phenotype
MIM: 150290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMC1 PubMed Links

Gene Details

Gene
LAMC1
Gene Name
laminin subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002293.3 Intron NP_002284.3

View Full Product Details