Product Details

SNP ID
rs7079
Assay Type
Functionally tested
NCBI dbSNP Submissions
74
Location
Chr.1:230702585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAAGGGAGAAATAACCAGCTAT[G/T]GTTCCGCATTCAAACAGAAATTCAG
Phenotype
MIM: 106150 MIM: 606974
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGT PubMed Links

Gene Details

Gene
AGT
Gene Name
angiotensinogen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000029.3 2522 UTR 3 NP_000020.1
Gene
COG2
Gene Name
component of oligomeric golgi complex 2
There are no transcripts associated with this gene.

View Full Product Details