Product Details

SNP ID
rs11819043
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:97767076 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCTTTCCTAAGGGGGGCTAGGATG[A/T]CGCGTACAGGGGCGGTGGGGAGAAC
Phenotype
MIM: 604158 MIM: 610243
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SFRP5 PubMed Links

Gene Details

Gene
SFRP5
Gene Name
secreted frizzled related protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003015.3 1558 UTR 3 NP_003006.2
Gene
ZFYVE27
Gene Name
zinc finger FYVE-type containing 27
There are no transcripts associated with this gene.

View Full Product Details