Product Details
- SNP ID
-
rs834996
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
63
- Location
-
Chr.1:99647180 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- GAAATCTGTGTCACAATCATTTATC[A/C]CTAGCTATAACTTTTGATGTAATTG
- Phenotype
-
MIM: 610182
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PALMD
PubMed Links
Gene Details
- Gene
- PALMD
- Gene Name
- palmdelphin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017734.4 |
|
Intron |
|
|
NP_060204.1 |
View Full Product Details