Product Details

SNP ID
rs10081966
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:161303870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGAAGGCAGATTGATTTTTTTT[A/T]AAAGACATTGAAGACACCCCTCAAA
Phenotype
MIM: 159440
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MPZ PubMed Links

Gene Details

Gene
MPZ
Gene Name
myelin protein zero
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000530.7 1181 Intron NP_000521.2
NM_001315491.1 1181 Intron NP_001302420.1
XM_017001321.1 1181 UTR 3 XP_016856810.1

View Full Product Details