Product Details

SNP ID
rs7639471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112336864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCATACTGCTAAATACCATAAAA[G/T]AATAGACAGAGCCAAATAAGAGACT
Phenotype
MIM: 155970
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD200 PubMed Links

Gene Details

Gene
CD200
Gene Name
CD200 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004196.3 Intron NP_001004196.2
NM_001318826.1 Intron NP_001305755.1
NM_001318828.1 Intron NP_001305757.1
NM_001318830.1 Intron NP_001305759.1
NM_005944.6 Intron NP_005935.4
XM_005247481.4 Intron XP_005247538.1
XM_005247482.2 Intron XP_005247539.1
XM_005247485.2 Intron XP_005247542.1
XM_005247486.2 Intron XP_005247543.1
XM_005247488.2 Intron XP_005247545.1
XM_017006452.1 Intron XP_016861941.1
XM_017006453.1 Intron XP_016861942.1
XM_017006454.1 Intron XP_016861943.1
XM_017006455.1 Intron XP_016861944.1
XM_017006456.1 Intron XP_016861945.1
XM_017006457.1 Intron XP_016861946.1
XM_017006458.1 Intron XP_016861947.1
XM_017006459.1 Intron XP_016861948.1
Gene
LOC105374042
Gene Name
uncharacterized LOC105374042
There are no transcripts associated with this gene.

View Full Product Details