Product Details
- SNP ID
-
rs10167187
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:191836667 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGCTCCTCCTTCCCTTCCACGGC[A/G]CCGGAAACGGGCTGTTTCACAAACA
- Phenotype
-
MIM: 606728
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SDPR
PubMed Links
Gene Details
- Gene
- SDPR
- Gene Name
- serum deprivation response
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004657.5 |
889 |
Silent Mutation |
GGC,GGT |
G178G |
NP_004648.1 |
View Full Product Details