Product Details

SNP ID
rs3810664
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:47447023 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAAAGTTTTTCCCAGTAGTGAGTA[C/T]TTTTATGTCCCAGGATATGCTGAAC
Phenotype
MIM: 314995
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF41 PubMed Links

Gene Details

Gene
ZNF41
Gene Name
zinc finger protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324139.1 3514 UTR 3 NP_001311068.1
NM_001324140.1 3514 UTR 3 NP_001311069.1
NM_001324141.1 3514 UTR 3 NP_001311070.1
NM_001324142.1 3514 UTR 3 NP_001311071.1
NM_001324143.1 3514 UTR 3 NP_001311072.1
NM_001324144.1 3514 UTR 3 NP_001311073.1
NM_001324145.1 3514 UTR 3 NP_001311074.1
NM_001324147.1 3514 UTR 3 NP_001311076.1
NM_001324148.1 3514 UTR 3 NP_001311077.1
NM_001324149.1 3514 UTR 3 NP_001311078.1
NM_001324150.1 3514 UTR 3 NP_001311079.1
NM_001324151.1 3514 UTR 3 NP_001311080.1
NM_001324152.1 3514 UTR 3 NP_001311081.1
NM_001324153.1 3514 UTR 3 NP_001311082.1
NM_001324154.1 3514 UTR 3 NP_001311083.1
NM_001324155.1 3514 UTR 3 NP_001311084.1
NM_001324156.1 3514 UTR 3 NP_001311085.1
NM_001324157.1 3514 UTR 3 NP_001311086.1
NM_007130.3 3514 UTR 3 NP_009061.1
NM_153380.3 3514 UTR 3 NP_700359.1
XM_006724550.3 3514 UTR 3 XP_006724613.1
XM_006724555.3 3514 UTR 3 XP_006724618.1
XM_017029810.1 3514 UTR 3 XP_016885299.1
XM_017029811.1 3514 UTR 3 XP_016885300.1
XM_017029812.1 3514 UTR 3 XP_016885301.1
XM_017029813.1 3514 UTR 3 XP_016885302.1
XM_017029814.1 3514 UTR 3 XP_016885303.1
XM_017029815.1 3514 UTR 3 XP_016885304.1
XM_017029816.1 3514 UTR 3 XP_016885305.1
XM_017029817.1 3514 UTR 3 XP_016885306.1

View Full Product Details