Product Details

SNP ID
rs2981011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128468584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAATTCCCTCCAGCCCCCTGCAG[C/G]CCTGCCTGGCCTGGGCAGCCCACCC
Phenotype
MIM: 611337
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB8 PubMed Links

Gene Details

Gene
DNAJB8
Gene Name
DnaJ heat shock protein family (Hsp40) member B8
There are no transcripts associated with this gene.

Gene
DNAJB8-AS1
Gene Name
DNAJB8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details