Product Details
- SNP ID
-
rs3747811
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:42271987 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACCAGTTGTATTTTTCTTCTCTCCC[A/T]TTCAAGAGCAGTGGTATCTCTTGCC
- Phenotype
-
MIM: 603258
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IKBKB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17875658] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IKBKB
- Gene Name
- inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
- Gene
- LOC101929897
- Gene Name
- uncharacterized LOC101929897
There are no transcripts associated with this gene.
View Full Product Details