Product Details

SNP ID
rs4738813
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:60683112 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAAAATCTGTTCTTTTCATGCGAT[C/T]GGAAATTCAGAATTTAGCATTTTTC
Phenotype
MIM: 608892
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHD7 PubMed Links

Gene Details

Gene
CHD7
Gene Name
chromodomain helicase DNA binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316690.1 Intron NP_001303619.1
NM_017780.3 Intron NP_060250.2
XM_011517553.2 Intron XP_011515855.1
XM_011517554.2 Intron XP_011515856.1
XM_011517555.2 Intron XP_011515857.1
XM_011517560.2 Intron XP_011515862.1
XM_017013612.1 Intron XP_016869101.1
XM_017013613.1 Intron XP_016869102.1

View Full Product Details