Product Details
- SNP ID
-
rs10468183
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:99971842 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGTACTCGAGGGACAGTACAGGGT[A/G]TTAACACAGAAACATCACTGCTGTG
- Phenotype
-
MIM: 607511
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAMTS17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs183671964] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAMTS17
- Gene Name
- ADAM metallopeptidase with thrombospondin type 1 motif 17
- Gene
- LOC105371022
- Gene Name
- uncharacterized LOC105371022
There are no transcripts associated with this gene.
View Full Product Details