Product Details

SNP ID
rs34805871
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:128886718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTAAAGGCTGAAGGTGGATCCA[C/T]AGAAGAACATGACAAACAACCAGGT
Phenotype
MIM: 156225
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LAMA2 PubMed Links

Gene Details

Gene
LAMA2
Gene Name
laminin subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000426.3 Intron NP_000417.2
NM_001079823.1 Intron NP_001073291.1
XM_005266981.3 Intron XP_005267038.1
XM_005266982.3 Intron XP_005267039.1
XM_011535820.2 Intron XP_011534122.1
XM_017010851.1 Intron XP_016866340.1
XM_017010852.1 Intron XP_016866341.1
XM_017010853.1 Intron XP_016866342.1

View Full Product Details