Product Details

SNP ID
rs4701195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179698397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGACTCTGCAGGAACTGCGGCGC[C/G]GGCTCACACAGCGCGGAGCAGTGCA
Phenotype
MIM: 114217
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CANX PubMed Links

Gene Details

Gene
CANX
Gene Name
calnexin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024649.1 Intron NP_001019820.1
NM_001746.3 Intron NP_001737.1
XM_011534664.1 Intron XP_011532966.1
XM_011534665.2 Intron XP_011532967.1

View Full Product Details