Product Details
- SNP ID
-
rs6581155
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:57784379 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGACGTTAAAGCACAGTACACATA[A/G]GCTACATTACATTTATAAGAAATTT
- Phenotype
-
MIM: 615258
MIM: 604723
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
METTL21B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs181907810] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- METTL21B
- Gene Name
- methyltransferase like 21B
There are no transcripts associated with this gene.
- Gene
- TSFM
- Gene Name
- Ts translation elongation factor, mitochondrial
View Full Product Details