Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001204171.1 | Intron | NP_001191100.1 | |||
NM_001204172.1 | Intron | NP_001191101.1 | |||
NM_001278516.1 | Intron | NP_001265445.1 | |||
NM_001278517.1 | Intron | NP_001265446.1 | |||
NM_001278518.1 | Intron | NP_001265447.1 | |||
NM_001278519.1 | Intron | NP_001265448.1 | |||
NM_002393.4 | Intron | NP_002384.2 | |||
XM_017001311.1 | Intron | XP_016856800.1 | |||
XM_017001312.1 | Intron | XP_016856801.1 | |||
XM_017001313.1 | Intron | XP_016856802.1 |