Product Details

SNP ID
rs7426919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46705485 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGTGGAGCCCAGGACTGGAGTA[A/G]GAGGGTGTGTTCATAGGGGCCTTGA
Phenotype
MIM: 607950 MIM: 607237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS50 PubMed Links

Gene Details

Gene
PRSS50
Gene Name
protease, serine 50
There are no transcripts associated with this gene.

Gene
TMIE
Gene Name
transmembrane inner ear
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147196.2 Intron NP_671729.2
XM_006713097.3 Intron XP_006713160.1

View Full Product Details