Product Details

SNP ID
rs3749212
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122368113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCAAAATTTTCATATTAAAAAT[C/T]TTACTTTGAAGGCTACAATTGGAAA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC58 PubMed Links

Gene Details

Gene
CCDC58
Gene Name
coiled-coil domain containing 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017928.3 Intron NP_001017928.1
NM_001308326.1 Intron NP_001295255.1
XM_011512410.2 Intron XP_011510712.1
XM_017005697.1 Intron XP_016861186.1

View Full Product Details