Product Details

SNP ID
rs4236040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25760981 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACTTGTTTTGGATGGATCACTT[C/T]TTTGAGTGTTTATAATTTTTTATTT
Phenotype
MIM: 182308 MIM: 604216
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC17A1 PubMed Links

Gene Details

Gene
SLC17A1
Gene Name
solute carrier family 17 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005074.3 Intron NP_005065.2
XM_011514818.2 Intron XP_011513120.2
XM_011514819.2 Intron XP_011513121.2
XM_011514820.2 Intron XP_011513122.2
XM_011514821.2 Intron XP_011513123.1
XM_017011199.1 Intron XP_016866688.1
XM_017011200.1 Intron XP_016866689.1
XM_017011201.1 Intron XP_016866690.1
XM_017011202.1 Intron XP_016866691.1
Gene
SLC17A4
Gene Name
solute carrier family 17 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286121.1 Intron NP_001273050.1
NM_005495.2 Intron NP_005486.1
XM_011514211.1 Intron XP_011512513.1
XM_011514213.2 Intron XP_011512515.1
XM_011514214.2 Intron XP_011512516.1
XM_011514215.2 Intron XP_011512517.1
XM_011514217.2 Intron XP_011512519.1
XM_011514218.2 Intron XP_011512520.1
XM_011514219.2 Intron XP_011512521.1
XM_017010150.1 Intron XP_016865639.1
XM_017010151.1 Intron XP_016865640.1

View Full Product Details