Product Details

SNP ID
rs36059541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:117680761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATAATTTGGGGTAAGCCTTA[A/G]GATTATTTGCTGTAAATTGGAATGA
Phenotype
MIM: 610463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927919 PubMed Links

Gene Details

Gene
LOC101927919
Gene Name
uncharacterized LOC101927919
There are no transcripts associated with this gene.

Gene
NUS1
Gene Name
NUS1 dehydrodolichyl diphosphate synthase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138459.3 Intron NP_612468.1

View Full Product Details