Product Details

SNP ID
rs3829944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36431153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTAATAACGCACACTTAGAAAAAC[G/T]GTGATAACCGTTGCTTGCATAAAGT
Phenotype
MIM: 604036
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNA1 PubMed Links

Gene Details

Gene
CCNA1
Gene Name
cyclin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111045.1 666 Intron NP_001104515.1
NM_001111046.1 666 Intron NP_001104516.1
NM_001111047.1 666 Intron NP_001104517.1
NM_003914.3 666 Intron NP_003905.1
XM_011535294.2 666 UTR 5 XP_011533596.1
XM_011535295.2 666 UTR 5 XP_011533597.1
XM_011535296.2 666 Intron XP_011533598.1

View Full Product Details