Product Details

SNP ID
rs3814803
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:36430827 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCGTTGGCCTCTGACTCTCTGTTGC[A/G]GCAATCTAGATGTTACCAGAATTCC
Phenotype
MIM: 604036
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNA1 PubMed Links

Gene Details

Gene
CCNA1
Gene Name
cyclin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111045.1 340 Intron NP_001104515.1
NM_001111046.1 340 Intron NP_001104516.1
NM_001111047.1 340 Intron NP_001104517.1
NM_003914.3 340 Intron NP_003905.1
XM_011535294.2 340 UTR 5 XP_011533596.1
XM_011535295.2 340 UTR 5 XP_011533597.1
XM_011535296.2 340 Intron XP_011533598.1

View Full Product Details