Product Details

SNP ID
rs11848612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104731230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCACTGTGCTTCTGAGAGTTGGG[C/G]GCTTACCTTAATCTACTGTCCCTGA
Phenotype
MIM: 612498
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADSSL1 PubMed Links

Gene Details

Gene
ADSSL1
Gene Name
adenylosuccinate synthase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320424.1 Intron NP_001307353.1
NM_152328.4 Intron NP_689541.1
NM_199165.2 Intron NP_954634.1
XM_006720026.3 Intron XP_006720089.1
XM_011536412.1 Intron XP_011534714.1
XM_011536414.2 Intron XP_011534716.1
Gene
LOC388022
Gene Name
uncharacterized LOC388022
There are no transcripts associated with this gene.

View Full Product Details