Product Details

SNP ID
rs10047036
Assay Type
Functionally tested
NCBI dbSNP Submissions
59
Location
Chr.1:53923527 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTAGTTCCCCTTATATGAATCTC[A/G]CTTGTCCTTTCATGCTTAAATTAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPB11 PubMed Links

Gene Details

Gene
HSPB11
Gene Name
heat shock protein family B (small) member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316935.1 Intron NP_001303864.1
NM_016126.3 Intron NP_057210.2

View Full Product Details