Product Details

SNP ID
rs1405209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99823263 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTAGAAATGAAAGTTCTCGGTGG[C/T]GAGACGAGAGCGGTCATGCGAGCGC
Phenotype
MIM: 600542
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928438 PubMed Links

Gene Details

Gene
LOC101928438
Gene Name
uncharacterized LOC101928438
There are no transcripts associated with this gene.

Gene
NR4A3
Gene Name
nuclear receptor subfamily 4 group A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006981.3 Intron NP_008912.2
NM_173199.2 Intron NP_775291.1
NM_173200.2 Intron NP_775292.1
XM_017015162.1 Intron XP_016870651.1

View Full Product Details