Product Details

SNP ID
rs17272966
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:53236705 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTGCCACATTTGTATGGTTTCCAT[C/T]CTGAATGTCTTCTGTTATAACCATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF677 PubMed Links

Gene Details

Gene
ZNF677
Gene Name
zinc finger protein 677
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317998.1 2434 UTR 3 NP_001304927.1
NM_182609.3 2434 UTR 3 NP_872415.1
XM_005258862.4 2434 UTR 3 XP_005258919.1
XM_011526912.2 2434 UTR 3 XP_011525214.1

View Full Product Details