Product Details
- SNP ID
-
rs34473043
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:86526904 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTATTTATTTAGGAATCTTAGTA[A/T]TGATTTTTGACATATGTTGAGGACT
- Phenotype
-
MIM: 604207
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE3
PubMed Links
Gene Details
- Gene
- CPNE3
- Gene Name
- copine 3
View Full Product Details