Product Details
- SNP ID
-
rs1540194
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:81888246 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTCCTTACAGCTGGTCTTTCTTTTC[A/G]TCTTAGACAATATCTACAGTGGAGT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4300
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114437963] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR4300
- Gene Name
- microRNA 4300
There are no transcripts associated with this gene.
- Gene
- MIR4300HG
- Gene Name
- MIR4300 host gene
There are no transcripts associated with this gene.
View Full Product Details