Product Details

SNP ID
rs11677037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61809520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCATCTCCGTTAGCATGTCTATGG[A/C]AGAGCTTTGATTCCTACCCACCCCA
Phenotype
MIM: 613596
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM161A PubMed Links

Gene Details

Gene
FAM161A
Gene Name
family with sequence similarity 161 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201543.1 Intron NP_001188472.1
NM_032180.2 Intron NP_115556.2
XM_017005072.1 Intron XP_016860561.1
XM_017005073.1 Intron XP_016860562.1
XM_017005074.1 Intron XP_016860563.1

View Full Product Details