Product Details
- SNP ID
-
rs35662666
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50309169 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCCTGAGCAGGCCCCCTGTGTG[C/T]ACCCTCAGCCCCGAGCCCCACTCCA
- Phenotype
-
MIM: 604293
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DENND6B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75807466] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DENND6B
- Gene Name
- DENN domain containing 6B
- Gene
- PLXNB2
- Gene Name
- plexin B2
There are no transcripts associated with this gene.
View Full Product Details