Product Details

SNP ID
rs13094785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50683478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTGTCCTAGAGGGGAATTATGT[C/T]TAGTGTATAAACAAGTAGTGTCTGC
Phenotype
MIM: 603123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK3 PubMed Links

Gene Details

Gene
DOCK3
Gene Name
dedicator of cytokinesis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004947.4 Intron NP_004938.1
XM_005264914.3 Intron XP_005264971.1
XM_005264915.3 Intron XP_005264972.1
XM_005264916.3 Intron XP_005264973.1
XM_005264917.3 Intron XP_005264974.1
XM_005264918.3 Intron XP_005264975.1
XM_006713008.3 Intron XP_006713071.1
XM_006713009.3 Intron XP_006713072.1
XM_006713010.3 Intron XP_006713073.1
XM_011533441.2 Intron XP_011531743.1
XM_011533443.2 Intron XP_011531745.1
XM_011533444.2 Intron XP_011531746.1
XM_011533445.2 Intron XP_011531747.1
XM_017005825.1 Intron XP_016861314.1
XM_017005826.1 Intron XP_016861315.1
XM_017005827.1 Intron XP_016861316.1
XM_017005828.1 Intron XP_016861317.1
Gene
MIR4787
Gene Name
microRNA 4787
There are no transcripts associated with this gene.

View Full Product Details