Product Details

SNP ID
rs4439944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233064066 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGTTGGCCGAGCCGGCATCCCC[C/T]TGTGCAATCTGCACAGGCAAAGCTG
Phenotype
MIM: 601582
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INPP5D PubMed Links

Gene Details

Gene
INPP5D
Gene Name
inositol polyphosphate-5-phosphatase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017915.2 Intron NP_001017915.1
NM_005541.4 Intron NP_005532.2
XM_017004004.1 Intron XP_016859493.1

View Full Product Details