Product Details
- SNP ID
-
rs10952
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:138252651 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCCTAATCTGGAATGCAATAGAGAA[A/T]GGCTAACTTATTAGATTCTGGTGAT
- Phenotype
-
MIM: 605710
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GFRA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112280020] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GFRA3
- Gene Name
- GDNF family receptor alpha 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001496.3 |
1660 |
UTR 3 |
|
|
NP_001487.2 |
View Full Product Details