Product Details

SNP ID
rs13213236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:129578006 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAGATGAATACTAACATTCTGTC[A/G]TGACAGGAATACTGTGAAATTAGGT
Phenotype
MIM: 613351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP18 PubMed Links

Gene Details

Gene
ARHGAP18
Gene Name
Rho GTPase activating protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033515.2 2463 UTR 3 NP_277050.2
XM_005267212.2 2463 UTR 3 XP_005267269.2
XM_005267213.1 2463 UTR 3 XP_005267270.1
XM_017011494.1 2463 UTR 3 XP_016866983.1

View Full Product Details