Product Details

SNP ID
rs8071254
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:17052770 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACATGGGCTACTGGCCTAGGCAGCA[G/A]CCACACCTTTGGCACAGCTCTGAAC
Phenotype
MIM: 612935
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MPRIP PubMed Links

Gene Details

Gene
MPRIP
Gene Name
myosin phosphatase Rho interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015134.3 Intron NP_055949.2
NM_201274.3 Intron NP_958431.2
XM_005256563.4 Intron XP_005256620.1
XM_005256564.4 Intron XP_005256621.3
XM_011523761.2 Intron XP_011522063.1
XM_011523762.2 Intron XP_011522064.1
XM_011523763.2 Intron XP_011522065.1
XM_011523764.2 Intron XP_011522066.1
XM_011523765.2 Intron XP_011522067.1
XM_011523766.2 Intron XP_011522068.2
XM_011523767.2 Intron XP_011522069.2
XM_017024393.1 Intron XP_016879882.1

View Full Product Details