Product Details

SNP ID
rs13206575
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:99921408 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTACTTACTTGGTGCATTGTGATA[C/T]ATTCTTTTTTATTCTTGTTGAAAAC
Phenotype
MIM: 606111
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCHR2 PubMed Links

Gene Details

Gene
MCHR2
Gene Name
melanin concentrating hormone receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040179.1 Intron NP_001035269.1
NM_032503.2 Intron NP_115892.2

View Full Product Details