Product Details

SNP ID
rs510399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168901407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTACCGAGCTTACTACACTTTT[C/T]TAAATTTTATGTCCAATGTTGGAGA
Phenotype
MIM: 608058
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
G6PC2 PubMed Links

Gene Details

Gene
G6PC2
Gene Name
glucose-6-phosphatase catalytic subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081686.1 168 Silent Mutation CTA,TTA L26L NP_001075155.1
NM_021176.2 168 Silent Mutation CTA,TTA L26L NP_066999.1
XM_011511564.2 168 Silent Mutation CTA,TTA L26L XP_011509866.1
XM_011511565.2 168 Intron XP_011509867.1

View Full Product Details