Product Details

SNP ID
rs721653
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:38876573 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACTCTTAGTTCCCATAGGCGTAGTC[T/C]TTCATGCCAAGTAAGAGCTCCAGGA
Phenotype
MIM: 615469
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
FAM114A1 PubMed Links

Gene Details

Gene
FAM114A1
Gene Name
family with sequence similarity 114 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138389.2 Intron NP_612398.2
XM_005262672.2 Intron XP_005262729.1
XM_006714033.2 Intron XP_006714096.1
XM_017008838.1 Intron XP_016864327.1
Gene
MIR574
Gene Name
microRNA 574
There are no transcripts associated with this gene.

View Full Product Details