Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001139514.1 | Intron | NP_001132986.1 | |||
NM_001139515.1 | Intron | NP_001132987.1 | |||
NM_053281.3 | Intron | NP_444511.1 | |||
XM_011530846.2 | Intron | XP_011529148.1 | |||
XM_011530847.2 | Intron | XP_011529149.1 | |||
XM_011530848.2 | Intron | XP_011529150.1 | |||
XM_017029254.1 | Intron | XP_016884743.1 | |||
XM_017029255.1 | Intron | XP_016884744.1 | |||
XM_017029256.1 | Intron | XP_016884745.1 |