Product Details

SNP ID
rs632015
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:63783617 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGTGAATCCTCCACCATGCCCA[A/G]AGCAGAATCCAGGCTAAAAGCCAGG
Phenotype
MIM: 602336
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ROR1 PubMed Links

Gene Details

Gene
ROR1
Gene Name
receptor tyrosine kinase like orphan receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083592.1 Intron NP_001077061.1
NM_005012.3 Intron NP_005003.2
XM_011541526.1 Intron XP_011539828.1
XM_017001376.1 Intron XP_016856865.1
XM_017001377.1 Intron XP_016856866.1

View Full Product Details