Product Details

SNP ID
rs525497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23666879 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGTAACCTCTGACCAACAACTAA[C/G]TCCCCATTCCTTCCCTCTCGCCTCG
Phenotype
MIM: 300927
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRDX4 PubMed Links

Gene Details

Gene
PRDX4
Gene Name
peroxiredoxin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006406.1 Intron NP_006397.1
XM_005274438.1 Intron XP_005274495.1
XM_017029230.1 Intron XP_016884719.1
XM_017029231.1 Intron XP_016884720.1

View Full Product Details