Product Details
- SNP ID
-
rs488282
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:106369238 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTATAGACAAGTAAAGGTTGCACCC[A/G]TAGCTAACACACACAAAAAGAAACA
- Phenotype
-
MIM: 601797
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AIM1
PubMed Links
Gene Details
- Gene
- AIM1
- Gene Name
- absent in melanoma 1
View Full Product Details