Product Details

SNP ID
rs553665
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:16550437 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCTATGTATGTTTTCCTCACTCCA[C/T]TAAAATTTCACCTTTTCCCCGACCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF287 PubMed Links

Gene Details

Gene
ZNF287
Gene Name
zinc finger protein 287
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020653.2 4158 UTR 3 NP_065704.2
XM_011523968.2 4158 UTR 3 XP_011522270.1
XM_017024887.1 4158 UTR 3 XP_016880376.1

View Full Product Details