Product Details

SNP ID
rs734192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:128269919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGGTGGGGGTGGGGGGCATGG[A/G]GGGACCCCGGGGAGCACAAAGCGCT
Phenotype
MIM: 604846
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS6ST1 PubMed Links

Gene Details

Gene
HS6ST1
Gene Name
heparan sulfate 6-O-sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004807.2 Intron NP_004798.3

View Full Product Details