Product Details

SNP ID
rs405567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:87933279 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCTATCAATGGCTGCCAACAAT[C/T]GGGAGTGAAAGGAACTGACTGAGCA
Phenotype
MIM: 606890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALC PubMed Links

Gene Details

Gene
GALC
Gene Name
galactosylceramidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000153.3 3900 UTR 3 NP_000144.2
NM_001201401.1 3900 UTR 3 NP_001188330.1
NM_001201402.1 3900 UTR 3 NP_001188331.1
XM_011536618.2 3900 UTR 3 XP_011534920.1

View Full Product Details