Product Details
- SNP ID
-
rs476851
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:8707324 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGTGGCCCGGGTTGAGGAGCAAC[A/G]GTGTTGCGCGGGCCAGTCCTTTGTA
- Phenotype
-
MIM: 616131
MIM: 615766
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GACAT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78564115] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GACAT2
- Gene Name
- gastric cancer associated transcript 2 (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- MTCL1
- Gene Name
- microtubule crosslinking factor 1
View Full Product Details