Product Details

SNP ID
rs450262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56998081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAAAGACTTTCCTCTCTTGTCC[A/C]AACACAAAAAAGCAAGCCAAGATCT
Phenotype
MIM: 605307
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR182 PubMed Links

Gene Details

Gene
GPR182
Gene Name
G protein-coupled receptor 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007264.3 2612 Intron NP_009195.1
XM_011537824.2 2612 UTR 3 XP_011536126.1
XM_011537825.2 2612 Intron XP_011536127.1
XM_011537827.2 2612 Intron XP_011536129.1
Gene
HBCBP
Gene Name
HBcAg-binding protein
There are no transcripts associated with this gene.

Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014830.2 2612 Intron NP_055645.1

View Full Product Details