Product Details
- SNP ID
-
rs1703874
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:614840 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTGTGGAGACTGATCTGAAAAAGCA[A/T]CCTTCGTGACAGGTGGAAGTTGGAT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ERICH1
PubMed Links
Gene Details
- Gene
- ERICH1
- Gene Name
- glutamate rich 1
View Full Product Details