Product Details

SNP ID
rs675759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:128838080 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAAACATTTTGATTCCTAAGCA[C/G]CTTTGAGTACCAGTTTTTTTCCAGG
Phenotype
MIM: 600359
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNJ1 PubMed Links

Gene Details

Gene
KCNJ1
Gene Name
potassium voltage-gated channel subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000220.4 2556 UTR 3 NP_000211.1
NM_153764.2 2556 UTR 3 NP_722448.1
NM_153765.2 2556 UTR 3 NP_722449.3
NM_153766.2 2556 UTR 3 NP_722450.1
NM_153767.3 2556 UTR 3 NP_722451.1

View Full Product Details